Order Entry
Denmark
Orders LinkContactUsLinkComponent
Tubidity standards for 2100A

Tubidity standards for 2100A

StabCal® standard calibration kit for model 2100A, 1,0 NTU, 100 ml bottles, 4× 100 ml, 1 KIT
Catalog # HACH2659110
Supplier:   Hach
Tubidity standards for 2100A
Catalog # HACH2659110
Supplier:  Hach
CAS Number:  
Tubidity standards for 2100A
Tubidity standards for 2100A

StabCal® standard calibration kit for model 2100A, 1,0 NTU, 100 ml bottles, 4× 100 ml, 1 KIT

CAS Number:  
Price
3.570,00 kr
Log in for availability
1 * 1 Each
Product Details & Documents

Stablcal Stabilised Formazin Turbidity Standards are true formazin dilutions developed for use in any turbidimeter.

  • Precise, pre-mixed formazin solutions
  • Based on Formazin Primary Standards
  • Enhanced stability saves time and labour
  • Confidence in results

Documents

Specifications

Specifications

  • Pack type:
    100 ml bottles
  • Pk:
    1 KIT
  • Turbidity:
    1,0 NTU
  • Description:
    StabCal® standard calibration kit for model 2100A
  • Pk Info:
    4× 100 ml

Specifications