SPE columns, CHROMABOND® C8
C8, 60 Å, 45 µm, 96×100 mg
Catalog # MANA738021.100M
Supplier: MACHEREY-NAGEL
Jump to:
Product Details & Documents
CHROMABOND® C8 is an octyl modified silica phase for SPE.
- Slightly more polar than C18 phase
- Hydrophobic and more pronounced secondary interactions with polar compounds
- Suitable for slight nonpolar and polar substances
Recommended for polar pesticides and PCBs.
Specifications
- Particle Size:45 µm
- Pore size:60 Å
- Seq:09
- Bed weight:96×100 mg
- Phase:C8