Order Entry
Denmark
Orders LinkContactUsLinkComponent
Tris-Borate-EDTA (TBE) buffer, 10X
Tris-Borate-EDTA (TBE) buffer, 10X
Tris-Borate-EDTA (TBE) buffer, 10X, Glass bottle, 1 L
Katalog # A0972.1000
Leverandør:   PanReac AppliChem
Tris-Borate-EDTA (TBE) buffer, 10X
Katalog # A0972.1000
Leverandør:  PanReac AppliChem
CAS nummer:  
Tris-Borate-EDTA (TBE) buffer, 10X
Tris-Borate-EDTA (TBE) buffer, 10X, Glass bottle, 1 L
CAS nummer:  
Price
566,00 kr
Log ind for tilgængelighed
1 * 1 L
Product Details & Documents
10X concentrated solution of 0,89 M Tris, 0,89 M boric acid and 0,02 M EDTA.Na₂, pH 8,3 ±0,2.
CT = Cubitainer®
Documents
Specifikationer
Specifikationer
  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Description:
    Tris-Borate-EDTA (TBE) buffer, 10X
Specifikationer
Anbefalinger