Order Entry
Denmark
Orders LinkContactUsLinkComponent
Buffer, PBS (1X, Dulbecco's) powder pack

Buffer, PBS (1X, Dulbecco's) powder pack

PBS buffer (1X, Dulbecco's), 50 l, 50 L
Catalog # A0964.9050
Supplier:   PanReac AppliChem
Buffer, PBS (1X, Dulbecco's) powder pack
Catalog # A0964.9050
Supplier:  PanReac AppliChem
CAS Number:  
Buffer, PBS (1X, Dulbecco's) powder pack
Buffer, PBS (1X, Dulbecco's) powder pack

PBS buffer (1X, Dulbecco's), 50 l, 50 L

CAS Number:  
Price
587,00 kr
Log in for availability
1 * 50 L
Product Details & Documents

Powder mixture to prepare PBS buffer (1X) according to Dulbecco.

Composed of di-sodium hydrogen phosphate anhydrous (1150 mg/l), potassium chloride (200 mg/l), potassium dihydrogen phosphate (200 mg/l) and sodium chloride (8000 mg/l).

Documents

Specifications

Specifications

  • Pk:
    50 L
  • Description:
    PBS buffer (1X, Dulbecco's), 50 l

Specifications