Order Entry
Denmark
Orders LinkContactUsLinkComponent
Sodium pyruvate

Sodium pyruvate

Glass bottle for solids, 100 g
Catalog # A4859.0100
Supplier:   PanReac AppliChem
Sodium pyruvate
Catalog # A4859.0100
Supplier:  PanReac AppliChem
CAS Number:  113-24-6
Sodium pyruvate

Glass bottle for solids, 100 g

CAS Number:  113-24-6
Price
1.070,00 kr
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  2-Ketopropionic acid sodium salt, 2-Oxopropionic acid sodium salt, Sodium 2-oxopropanoate, Pyruvic acid sodium salt, α-Ketopropionic acid sodium salt
  • CAS Number:113-24-6
  • Formula:CH₃COCOONa
  • MDL Number:MFCD00002586
  • Molecular Weight:110.04 g/mol
  • Melting Pt:220…230 °C
  • Density:1.718 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:204-024-4

Documents

Certificates
Search by Lot Number
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications