Order Entry
Denmark
Orders LinkContactUsLinkComponent
Colchicine

Colchicine

Glass bottle for solids, 5 g
Catalog # A4082.0005
Toxic
Health
Supplier:   PanReac AppliChem
Colchicine
Catalog # A4082.0005
Supplier:  PanReac AppliChem
CAS Number:  64-86-8
Colchicine

Glass bottle for solids, 5 g

CAS Number:  64-86-8
Price
4.080,00 kr
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  7α-H-Colchicine, (S)-(-)-Colchicine
  • CAS Number:64-86-8
  • Formula:C₂₂H₂₅NO₆
  • MDL Number:MFCD00078484
  • Molecular Weight:399.44 g/mol
  • Merck Index:12,02536
  • Melting Pt:154…156 °C
  • Density:1.32 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-598-5
  • UN:1544
  • ADR:6.1,I

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications