Single Full Rack holds (4) 3 Boxes 5 1/2 W x 5 1/2 D x 13 1/8 H (old style rack) 1 * 1 items
REVC5981
Single Full Rack holds (4) 3 Boxes 5 1/2 W x 5 1/2 D x 13 1/8 H (old style rack) 1 * 1 items
REVC5981
Frequently Bought Together
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR® Serological Pipette, Standard Line
: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR® Facial Tissues
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection