TRIS-TAPS-EDTA (TTE) BUFFER (TTE Buffer for DNA Sequencing) 1 * 1 l
Katalog # K370-1L
Leverandør: VWR Chemicals
TRIS-TAPS-EDTA (TTE) BUFFER (TTE Buffer for DNA Sequencing) 1 * 1 l
Katalog # K370-1L
Leverandør: VWR Chemicals
Frequently Bought Together
Vand, HiPerSolv CHROMANORM® for HPLC
Leverandør: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Leverandør: VWR Chemicals
Sealing film for manual application, Parafilm® M
Leverandør: HEATHROW
Acetone ≥99%, TECHNICAL
Leverandør: VWR Chemicals
pH-teststrimler, MQuant®, Supelco®
Leverandør: Merck
VWR®, Ansigtsservietter
Leverandør: VWR Collection
Acetonitril ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Leverandør: VWR Chemicals
VWR®, Nitril handsker, NITRILE
Leverandør: VWR Collection
Handsker, nitril, Kimtech™ Purple Nitrile™
Leverandør: KIMBERLY CLARK
VWR®, Serologiske pipetter, Standard type
Leverandør: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
Leverandør: VWR Collection
VWR®, Transfer Pipettes, High Performance
Leverandør: VWR Collection