Order Entry
Denmark
Orders LinkContactUsLinkComponent
VWR®, Yeast extract
VWR®, Yeast extract
Yeast extract, 100 g
Catalog # J850-100G
Supplier:   VWR Chemicals
VWR®, Yeast extract
Catalog # J850-100G
Supplier:  VWR Chemicals
CAS Number:  
VWR®, Yeast extract
VWR®, Yeast extract
Yeast extract, 100 g
CAS Number:  
Price
875,00 kr
Log in for availability
1 * 100 g
Product Details & Documents
Yeast extract is the water soluble portion of autolysed yeast containing a source of Vitamin B complex.
  • High purity extract
  • Used for supplementing media
Specifications
Specifications
  • Pk:
    100 g
  • Description:
    Yeast extract
Specifications