Order Entry
Denmark
Orders LinkContactUsLinkComponent
D-(-)-Fructose ≥99%, high purity

D-(-)-Fructose ≥99%, high purity

Plastic bucket, 5 kg
Catalog # 0226-5KG
Supplier:   VWR Chemicals
D-(-)-Fructose ≥99%, high purity
Catalog # 0226-5KG
Supplier:  VWR Chemicals
CAS Number:  57-48-7
D-(-)-Fructose ≥99%, high purity
D-(-)-Fructose ≥99%, high purity

Plastic bucket, 5 kg

CAS Number:  57-48-7
Price
2.030,00 kr
Log in for availability
1 * 5 kg
Product Details & Documents
Synonyms:  D(-)-Levulose, (3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one, β-D-(-)-Fructose, Fructose
  • CAS Number:57-48-7
  • Formula:C₆H₁₂O₆
  • MDL Number:MFCD00148910
  • Molecular Weight:180.16 g/mol
  • Merck Index:13,04295
  • Boiling Pt:552 °C (1013 hPa)
  • Melting Pt:100…110 °C
  • Density:1.59 g/cm³ (20 °C. OECD 109)
  • Storage Temperature:Ambient
  • EINECS:200-333-3
Specifications

Specification Test Results

  • Arsenic:
    <= 0.0001%
  • Calcium & Magnesium:
    <= 0.005%
  • Chloride:
    <= 0.018%
  • Heavy Metals (as Pb):
    <= 0.0005%
  • Loss on Drying:
    <= 0.5%
  • Purity:
    >= 99%
  • Residue on Ignition:
    <= 0.5%
  • Sulphate:
    <= 0.025%

Specifications

  • Pack type:
    Plastic bucket
  • Pk:
    5 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.