Order Entry
Denmark
Orders LinkContactUsLinkComponent
D-(-)-Fructose ≥99%, high purity

D-(-)-Fructose ≥99%, high purity

Plastic bottle, 1 kg
Catalog # 0226-1KG
Supplier:   VWR Chemicals
D-(-)-Fructose ≥99%, high purity
Catalog # 0226-1KG
Supplier:  VWR Chemicals
CAS Number:  57-48-7
D-(-)-Fructose ≥99%, high purity
D-(-)-Fructose ≥99%, high purity

Plastic bottle, 1 kg

CAS Number:  57-48-7
Price
698,00 kr
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  D(-)-Levulose, (3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one, β-D-(-)-Fructose, Fructose
  • CAS Number:57-48-7
  • Formula:C₆H₁₂O₆
  • MDL Number:MFCD00148910
  • Molecular Weight:180.16 g/mol
  • Merck Index:13,04295
  • Boiling Pt:552 °C (1013 hPa)
  • Melting Pt:100…110 °C
  • Density:1.59 g/cm³ (20 °C. OECD 109)
  • Storage Temperature:Ambient
  • EINECS:200-333-3
Specifications

Specification Test Results

  • Arsenic:
    <= 0.0001%
  • Calcium & Magnesium:
    <= 0.005%
  • Chloride:
    <= 0.018%
  • Heavy Metals (as Pb):
    <= 0.0005%
  • Loss on Drying:
    <= 0.5%
  • Purity:
    >= 99%
  • Residue on Ignition:
    <= 0.5%
  • Sulphate:
    <= 0.025%

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 kg

Specifications