Spray bottles
850 ml
Catalog # 215-0110
Supplier: Brand
Jump to:
Product Details & Documents
HDPE, white.
- Good chemical resistance
- Good temperature resistance
- Robust, convenient, stable
By turning the nozzle, delivery can be varied from a fine spray to a solid stream.
Specifications
- Capacity:850 ml
- Seq:03