Order Entry
Denmark
Orders LinkContactUsLinkComponent
Inoculating loops with needle end
Inoculating loops with needle end
Inoculating loops, flexible, 1 µl, Natural, 50 bags with 20 pieces
Catalog # 631-2216
Supplier:   Brand
Inoculating loops with needle end
Catalog # 631-2216
Supplier:  Brand
Inoculating loops with needle end
Inoculating loops with needle end
Inoculating loops, flexible, 1 µl, Natural, 50 bags with 20 pieces
Price
771,00 kr
Log in for availability
1 * 1.000 Items
Product Details & Documents
PS, gamma sterilised, disposable. For inoculation of nutrient media. Available either with loop at one end and needle at the other or loops at both ends. High flexibility permits gentle streaking without damaging the nutrient surface.
Specifications
  • Packed:
    50 bags with 20 pieces
  • Capacity:
    1 µl
  • Colour:
    Natural
  • Description:
    Inoculating loops, flexible