Dropper tip caps, WHEATON®
15-415, Red
Catalog # WHEAW242512-A
Jump to:
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
Cap,dropper tip,15-415,pp red 1 * 100 items Catalog #215-2764
Produktet 215-2764 er en erstatning for WHEAW242512-A, men varierer efter mærke, emballage eller mængde.
Product Details & Documents
Screw cap made of radiation grade polypropylene.
- Very good chemical resistance
- Very good temperature resistance
- Variety of colors
Specifications
- Thread:15-415
- Seq:33
- Cap colour:Red