Order Entry
Denmark
Orders LinkContactUsLinkComponent
Dropper tip caps, WHEATON®
Dropper tip caps, WHEATON®
20-410, Red
Catalog # WHEAW242522-A
Supplier:   DWK Life Sciences
Dropper tip caps, WHEATON®
Catalog # WHEAW242522-A
Supplier:  DWK Life Sciences
Dropper tip caps, WHEATON®
Dropper tip caps, WHEATON®
20-410, Red
Price
4.870,00 kr
Log in for availability
1 * 1.000 Items
Product Details & Documents
Screw cap made of radiation grade polypropylene.
  • Very good chemical resistance
  • Very good temperature resistance
  • Variety of colors
Specifications
  • Thread:
    20-410
  • Seq:
    47
  • Cap colour:
    Red