Order Entry
Denmark
Orders LinkContactUsLinkComponent
Dropper tip caps, WHEATON®
Dropper tip caps, WHEATON®
8-425, Yellow
Katalog # 215-4040
Leverandør:   DWK Life Sciences
Dropper tip caps, WHEATON®
Katalog # 215-4040
Leverandør:  DWK Life Sciences
Dropper tip caps, WHEATON®
Dropper tip caps, WHEATON®
8-425, Yellow
Price
3.840,00 kr
Log ind for tilgængelighed
1 * 1.000 Items
Product Details & Documents
Screw cap made of radiation grade polypropylene.
  • Very good chemical resistance
  • Very good temperature resistance
  • Variety of colors
Specifikationer
  • Thread:
    8-425
  • Seq:
    13
  • Cap colour:
    Yellow
Anbefalinger