BOD bottles, WHEATON®
BOD bottles, with bar codes, borosilicate glass, 69×165 mm (Ø×H), number sequence: 169-192, 300 ml
Catalog # WHEA227497-08
Supplier: DWK Life Sciences
Jump to:
Product Details & Documents
Made of Type I borosilicate glass.
- Available with or without stopper
- With white labeling area
Specifications
- Kapacitet:300 ml
- Beskrivelse:BOD bottles, with bar codes, borosilicate glass, 69×165 mm (Ø×H), number sequence: 169-192
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



