Order Entry
Denmark
Orders LinkContactUsLinkComponent
BOD bottles, WHEATON®
BOD bottles, WHEATON®
BOD bottles, with bar codes, borosilicate glass, 69×165 mm (Ø×H), number sequence: 97-120, 300 ml
  WHEA227497-05
BOD bottles, WHEATON®
  WHEA227497-05
BOD bottles, WHEATON®
BOD bottles, WHEATON®
BOD bottles, with bar codes, borosilicate glass, 69×165 mm (Ø×H), number sequence: 97-120, 300 ml
6.710,00 kr
1 * 24 Items
Made of Type I borosilicate glass.
  • Available with or without stopper
  • With white labeling area
  • Capacity:
    300 ml
  • Description:
    BOD bottles, with bar codes, borosilicate glass, 69×165 mm (Ø×H), number sequence: 97-120