Order Entry
Denmark
Orders LinkContactUsLinkComponent
BOD bottles, WHEATON®
BOD bottles, WHEATON®
BOD bottles, borosilicate glass, 43×115 mm (Ø×H), number sequence: 73-108, 60 ml
Catalog # WHEA227494-03G
Supplier:   DWK Life Sciences
BOD bottles, WHEATON®
Catalog # WHEA227494-03G
Supplier:  DWK Life Sciences
BOD bottles, WHEATON®
BOD bottles, WHEATON®
BOD bottles, borosilicate glass, 43×115 mm (Ø×H), number sequence: 73-108, 60 ml
Price
19.480,00 kr
Log in for availability
1 * 36 Items
Product Details & Documents
Made of Type I borosilicate glass.
  • Available with or without stopper
  • With white labeling area
Specifications
  • Capacity:
    60 ml
  • Description:
    BOD bottles, borosilicate glass, 43×115 mm (Ø×H), number sequence: 73-108
Anbefalinger