Dropper tip caps, WHEATON®
13-425, Blue
Catalog # 215-4031
Supplier: DWK Life Sciences
Jump to:
Product Details & Documents
Screw cap made of radiation grade polypropylene.
- Very good chemical resistance
- Very good temperature resistance
- Variety of colors
Specifications
- Thread:13-425
- Seq:18
- Cap colour:Blue