Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

Saccharometers

30 - 40, 0,1, 260 mm
  AMARH845034
 :   Amarell
Saccharometers
  AMARH845034
 :  Amarell
Saccharometers
Saccharometers

30 - 40, 0,1, 260 mm

 
272,00 kr
   
1 * 1 Items
 

Saccharometers with reference temperature of 20 °C.

 
  • Length:
    260 mm
  • Division:
    0,1
  • Range:
    30 - 40