Manikins
Manikin Baby Care Model Female
Catalog # BINH1005089
Marketsource
Supplier: 3B Scientific
Jump to:
Product Details & Documents
This full-size manikin allows the training of BLS rescue techniques and CPR. The resiliency and weight of the body is very realistic.
- Several models
Specifications
- Description:Manikin Baby Care Model Female