Order Entry
Denmark
Orders LinkContactUsLinkComponent
Filter papers, reusable, Aesculap®
Filter papers, reusable, Aesculap®
171×231 mm
Catalog # AESCJK092
Supplier:   Aesculap
Filter papers, reusable, Aesculap®
Catalog # AESCJK092
Supplier:  Aesculap
Filter papers, reusable, Aesculap®
171×231 mm
Price
Inquire For Price
Log in for availability
1 * 2 Items
Product Details & Documents
Reusable filters for 1000 cleaning and sterilisation cycles.
Specifications
  • Seq:
    02
  • Size:
    171×231 mm