Dilator set, Aesculap®
Dilator, 26 mm
Catalog # AESCEM126R
Supplier: Aesculap
Jump to:
Product Details & Documents
Complete set of uterine dilators.
Delivery information: Consists of 26 dilators, 1 rack for dilators (1 to 17 mm), 1 rack for dilators (18 to 26 mm) and metal case with hinged aluminium lid (330×220×80 mm).
Specifications
- Description:Dilator, 26 mm
- Seq:27
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



