Order Entry
Denmark
Orders LinkContactUsLinkComponent
Tissue grasping forceps, Aesculap®

Tissue grasping forceps, Aesculap®

Tissue grasping forceps, type Braun, 4×4 teeth (10 mm), 155 mm
Catalog # AESCBF311R
Supplier:   Aesculap
Tissue grasping forceps, Aesculap®
Catalog # AESCBF311R
Supplier:  Aesculap
Tissue grasping forceps, Aesculap®

Tissue grasping forceps, type Braun, 4×4 teeth (10 mm), 155 mm

Price
Inquire For Price
Log in for availability
1 * 1 Items
Product Details & Documents
Specifications
  • Description:
    Tissue grasping forceps, type Braun, 4×4 teeth (10 mm)
  • Length:
    155 mm
  • Seq:
    03
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.