Order Entry
Denmark
Orders LinkContactUsLinkComponent
Tissue forceps, Aesculap®
Tissue forceps, Aesculap®
Tissue forceps, 3×4 teeth, Straight, 200 mm
Catalog # AESCBD591R
Supplier:   Aesculap
Tissue forceps, Aesculap®
Catalog # AESCBD591R
Supplier:  Aesculap
Tissue forceps, Aesculap®
Tissue forceps, 3×4 teeth, Straight, 200 mm
Price
Inquire For Price
Log in for availability
1 * 1 Items
Product Details & Documents
For use in surgery.
Specifications
  • Type:
    Straight
  • Description:
    Tissue forceps, 3×4 teeth
  • Length:
    200 mm
  • Seq:
    06