Order Entry
Denmark
Orders LinkContactUsLinkComponent
Laboratory scissors, straight
Laboratory scissors, straight
Sharp/blunt, 150 mm
Catalog # BOCH4004
Supplier:   BOCHEM
Laboratory scissors, straight
Catalog # BOCH4004
Supplier:  BOCHEM
Laboratory scissors, straight
Laboratory scissors, straight
Sharp/blunt, 150 mm
Price
103,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Straight scissors made of stainless steel.
Caution: These scissors are not recommended for autoclaving.
Documents
Specifications
  • Type:
    Sharp/blunt
  • Length:
    150 mm
  • Seq:
    05