Order Entry
Denmark
Orders LinkContactUsLinkComponent
Beakers, low form

Beakers, low form

1000 ml, 130 mm, 100 mm
Catalog # BOCH8503
Supplier:   BOCHEM
Beakers, low form
Catalog # BOCH8503
Supplier:  BOCHEM
Beakers, low form
Beakers, low form

1000 ml, 130 mm, 100 mm

Price
1.030,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents

Stainless steel, 18/10.

  • Good chemical resistance
  • Excellent resistance to heat
  • Smooth surfaces for fast, easy cleaning

Ideal for corrosive materials. Robust and durable.

Specifications
  • Ext.Ø:
    100 mm
  • Capacity:
    1000 ml
  • Height:
    130 mm