Order Entry
Denmark
Orders LinkContactUsLinkComponent
Beakers, low form

Beakers, low form

250 ml, 85 mm, 65 mm
Catalog # BOCH8501
Supplier:   BOCHEM
Beakers, low form
Catalog # BOCH8501
Supplier:  BOCHEM
Beakers, low form
Beakers, low form

250 ml, 85 mm, 65 mm

Price
640,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

Stainless steel, 18/10.

  • Good chemical resistance
  • Excellent resistance to heat
  • Smooth surfaces for fast, easy cleaning

Ideal for corrosive materials. Robust and durable.

Specifikationer
  • Ext.Ø:
    65 mm
  • Capacity:
    250 ml
  • Height:
    85 mm