Order Entry
Denmark
Orders LinkContactUsLinkComponent
Beakers, low form

Beakers, low form

1000 ml, 130 mm, 100 mm
Catalog # BOCH8523
Supplier:   BOCHEM
Beakers, low form
Catalog # BOCH8523
Supplier:  BOCHEM
Beakers, low form
Beakers, low form

1000 ml, 130 mm, 100 mm

Price
1.270,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents

Stainless steel, 18/10.

  • Good chemical resistance
  • Excellent resistance to heat
  • Smooth surfaces for fast, easy cleaning

Ideal for corrosive materials. Robust and durable.

Specifications
  • Ext.Ø:
    100 mm
  • Capacity:
    1000 ml
  • Height:
    130 mm
Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.