Order Entry
Denmark
Orders LinkContactUsLinkComponent
Retort rods, with M10 thread

Retort rods, with M10 thread

16 mm, 1250 mm, M10
Catalog # BOCH5117
Supplier:   BOCHEM
Retort rods, with M10 thread
Catalog # BOCH5117
Supplier:  BOCHEM
Retort rods, with M10 thread
Retort rods, with M10 thread

16 mm, 1250 mm, M10

Price
794,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents

Stainless steel 18/10.

  • Rods for stand bases
  • M10 thread
Specifications
  • Ø:
    16 mm
  • Thread:
    M10
  • Length:
    1250 mm
  • Seq:
    08