Reading pocket magnifiers
Folding magnifier in black leather case, 60 mm
ESCH1752560
These handheld folding magnifiers with classic design fit easily into any handbag or pocket.
- Many different colours and sizes
- Anti-scratch coating for both models
- Black or brown leather case available
- Ø:60 mm
- Magnification:3,5×
- Description:Folding magnifier in black leather case