Order Entry
Denmark
ContactUsLinkComponent
Medical/Pharmaceutical Refrigerator. 158 litre interior capacity. Electronic temperature control (range 2C to 14C) with digital di splay, Two lockable double glazed sliding glass doors and heat reflective film. 1 * 1 items
  472-1123
Medical/Pharmaceutical Refrigerator. 158 litre interior capacity. Electronic temperature control (range 2C to 14C) with digital di splay, Two lockable double glazed sliding glass doors and heat reflective film. 1 * 1 items
  472-1123
Frequently Bought Together