Order Entry
Denmark
Orders LinkContactUsLinkComponent
Hydrophobic interaction chromatography columns, TSK-GEL®

Hydrophobic interaction chromatography columns, TSK-GEL®

Ether-5PW, Analytical, Ether-5PW, 1000 Å, 10 µm, 7,50 mm, 75 mm
Catalog # TOYO0008641
Supplier:   TOSOH Bioscience
Hydrophobic interaction chromatography columns, TSK-GEL®
Catalog # TOYO0008641
Supplier:  TOSOH Bioscience
Hydrophobic interaction chromatography columns, TSK-GEL®
Hydrophobic interaction chromatography columns, TSK-GEL®

Ether-5PW, Analytical, Ether-5PW, 1000 Å, 10 µm, 7,50 mm, 75 mm

Price
14.850,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents

These are polymethacrylate-based columns with a choice of three ligands (ether, phenyl or butyl) with varied hydrophobicities from low to high. Suitable for LC-MS applications.

  • Rigid polymeric base resin
  • Eliminates undesirable interactions with column hardware
  • Wide pH range (2 to 12) enables robust cleaning options

 

Specifications
  • Column Format:
    Analytical
  • Ø int.:
    7,50 mm
  • Particle Size:
    10 µm
  • Pore size:
    1000 Å
  • Description:
    Ether-5PW
  • Length:
    75 mm
  • Phase:
    Ether-5PW
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.