Order Entry
Denmark
Orders LinkContactUsLinkComponent
Capillary columns for ASTM method

Capillary columns for ASTM method

Columns for analysing PCBs in insulating liquids, Capillary, 0,25 µm, 0,25 mm, 50 m
Catalog # VARICP7482
Supplier:   VARIAN
Capillary columns for ASTM method
Catalog # VARICP7482
Supplier:  VARIAN
Capillary columns for ASTM method

Columns for analysing PCBs in insulating liquids, Capillary, 0,25 µm, 0,25 mm, 50 m

Price
14.490,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Specifications
  • Column Format:
    Capillary
  • Film thickness:
    0,25 µm
  • Ø int.:
    0,25 mm
  • Description:
    Columns for analysing PCBs in insulating liquids
  • Length:
    50 m
  • Seq:
    01