Order Entry
Denmark
Orders LinkContactUsLinkComponent
Instrument trays, rectangular
Instrument trays, rectangular
280×425×50 mm
Catalog # AGEN1242
Supplier:   Agencinox
Instrument trays, rectangular
Catalog # AGEN1242
Supplier:  Agencinox
Instrument trays, rectangular
280×425×50 mm
Price
821,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Stainless steel, 18/10.
  • Smooth surfaces for fast, easy cleaning
  • Good chemical resistance
  • Excellent thermal resistance
Specifications
  • Seq:
    01
  • W×D×H:
    280×425×50 mm