Order Entry
Denmark
Orders LinkContactUsLinkComponent
pH Indicator solution, wide range pH 4 - 10

pH Indicator solution, wide range pH 4 - 10

Plastic bottle, 500 mL
Catalog # HACH2329649
Supplier:   Hach
pH Indicator solution, wide range pH 4 - 10
Catalog # HACH2329649
Supplier:  Hach
CAS Number:  
pH Indicator solution, wide range pH 4 - 10
pH Indicator solution, wide range pH 4 - 10

Plastic bottle, 500 mL

CAS Number:  
Price
388,00 kr
Log in for availability
1 * 500 mL
Product Details & Documents

Wide range of pH indicator solutions used for visual determination of pH, using test kits.

Method:10222

Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.