Order Entry
Denmark
Orders LinkContactUsLinkComponent
Pipette tips
1000 - 5000 µl, +, Transparent, Oxford-Gilson automatic pipettes
  KIMA18266
Pipette tips
  KIMA18266
Pipette tips
1000 - 5000 µl, +, Transparent, Oxford-Gilson automatic pipettes
173,00 kr
1 * 300 Items
  • CE-IVD:
    +
  • Volume:
    1000 - 5000 µl
  • Tip colour:
    Transparent
  • For use with:
    Oxford-Gilson automatic pipettes