Order Entry
Denmark
Orders LinkContactUsLinkComponent
Transfer pipettes, fine tip

Transfer pipettes, fine tip

Extended fine tip, standard, 3,3 ml, 20 µl, 157 mm, +, Bulk
Catalog # 612-0006
Supplier:   Alpha Laboratories
Transfer pipettes, fine tip
Catalog # 612-0006
Supplier:  Alpha Laboratories
Transfer pipettes, fine tip
Transfer pipettes, fine tip

Extended fine tip, standard, 3,3 ml, 20 µl, 157 mm, +, Bulk

Price
1.930,00 kr
Log in for availability
1 * 500 Items
Product Details & Documents

Non toxic, food grade LDPE.

  • For careful control and small volume transfers
  • Ideal for supernatant removal, slide preparation and gel loading
Specifications
  • Packed:
    Bulk
  • Sterile:
    +
  • Capacity:
    3,3 ml
  • Description:
    Extended fine tip, standard
  • Length:
    157 mm
  • Seq:
    08
  • Drop vol.:
    20 µl
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.