Order Entry
Denmark
Orders LinkContactUsLinkComponent
Transfer pipettes, fine tip

Transfer pipettes, fine tip

Extended fine tip, mini, 1,0 ml, 20 µl, 104 mm, -, Bulk
Catalog # 612-0005
Supplier:   Alpha Laboratories
Transfer pipettes, fine tip
Catalog # 612-0005
Supplier:  Alpha Laboratories
Transfer pipettes, fine tip
Transfer pipettes, fine tip

Extended fine tip, mini, 1,0 ml, 20 µl, 104 mm, -, Bulk

Price
1.070,00 kr
Log in for availability
1 * 400 Items
Product Details & Documents

Non toxic, food grade LDPE.

  • For careful control and small volume transfers
  • Ideal for supernatant removal, slide preparation and gel loading
Specifications
  • Packed:
    Bulk
  • Sterile:
    -
  • Capacity:
    1,0 ml
  • Description:
    Extended fine tip, mini
  • Length:
    104 mm
  • Seq:
    03
  • Drop vol.:
    20 µl