Order Entry
Denmark
Orders LinkContactUsLinkComponent
Retort rods, with M10 thread

Retort rods, with M10 thread

13 mm, 1000 mm, M10
Catalog # BOCH5173
Supplier:   BOCHEM
Retort rods, with M10 thread
Catalog # BOCH5173
Supplier:  BOCHEM
Retort rods, with M10 thread
Retort rods, with M10 thread

13 mm, 1000 mm, M10

Price
357,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents

Stainless steel 18/10.

  • Rods for stand bases
  • M10 thread
Specifications
  • Ø:
    13 mm
  • Thread:
    M10
  • Length:
    1000 mm
  • Seq:
    13
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.