Order Entry
Denmark
Orders LinkContactUsLinkComponent
Clamps
Clamps
40 mm
Catalog # 241-7189
Supplier:   BOCHEM
Clamps
Catalog # 241-7189
Supplier:  BOCHEM
Clamps
Clamps
40 mm
Price
157,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Malleable cast iron, chromed.
  • Jaws have cork inserts to protect clamped items
  • Without bosshead for fixing to stand
  • Wing screws for rapid, easy clamping
DIN 12894
Certifications: DIN 12894
Specifications
  • Spanning width:
    40 mm