Protease inhibitor cocktail set v edta-free,10 vials 1 * 10 Vial
Catalog # 539137-10
Supplier: Merck Millipore (Calbiochem)
Protease inhibitor cocktail set v edta-free,10 vials 1 * 10 Vial
Catalog # 539137-10
Supplier: Merck Millipore (Calbiochem)
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR® Facial Tissues
Supplier: VWR Collection
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
PROTEASE INHIBITOR COCKTAIL SET V, EDTA-FREE 1 * 1 mL
Supplier: Merck Millipore (Calbiochem)
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Round-bottom polystyrene tubes, disposable, Falcon®
Supplier: Corning
VWR® PCR Tubes, 0,2 ml
Supplier: VWR Collection
X-Gal (5-bromo-4-chloro-3-indolyl-ß-D-galactopyranoside)
Supplier: PanReac AppliChem
Serum-free, haematopoietic media, X-VIVO™
Supplier: LONZA