Order Entry
Denmark
Orders LinkContactUsLinkComponent
Serrefines
Serrefines
Serrefine, Straight, 50 mm
Catalog # 221-0053
Supplier:   HAMMACHER KARL
Serrefines
Catalog # 221-0053
Supplier:  HAMMACHER KARL
Serrefines
Serrefines
Serrefine, Straight, 50 mm
Price
764,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
These small forceps for clamping a blood vessel are made from stainless steel.
Specifications
  • Type:
    Straight
  • Description:
    Serrefine
  • Length:
    50 mm
  • Seq:
    05