Order Entry
Denmark
Orders LinkContactUsLinkComponent
Wire baskets, autoclavable, Poxygrid®
Wire baskets, autoclavable, Poxygrid®
630×360×150 mm
Catalog # BELAH167590000
Wire baskets, autoclavable, Poxygrid®
Catalog # BELAH167590000
Wire baskets, autoclavable, Poxygrid®
Wire baskets, autoclavable, Poxygrid®
630×360×150 mm
Price
2.360,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
These epoxy-coated, steel wire baskets are suitable for transporting and batch dipping glassware.
  • Resistant to rust, corrosion, solvents, acids and alkalis
  • Withstands a temperature range of −223 to +121 °C
Specifications
  • W×D×H:
    630×360×150 mm