Order Entry
Denmark
Orders LinkContactUsLinkComponent
Test strips

Test strips

pH, pH 4 - 9, TS, 50
Catalog # 705-0189
Supplier:   Hach
Test strips
Catalog # 705-0189
Supplier:  Hach
CAS Number:  
Test strips
Test strips

pH, pH 4 - 9, TS, 50

CAS Number:  
Price
143,00 kr
Log in for availability
1 * 50 Items
Product Details & Documents

Simple to handle test strips give an overview of the water quality in just a few seconds. In the laboratory the strips can be used as an initial screening to indicate measuring range or potential problems.

  • Portable - ideal for field testing of surface water
  • Easy to use

Documents

Specifications

Specifications

  • Pk:
    50
  • Description:
    pH
  • Type:
    TS
  • Range:
    pH 4 - 9

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.