Order Entry
Denmark
Orders LinkContactUsLinkComponent
Tissue processing cassettes, Histosette I

Tissue processing cassettes, Histosette I

White
Catalog # DELT454141
Supplier:   DELTALAB
Tissue processing cassettes, Histosette I
Catalog # DELT454141
Supplier:  DELTALAB
Tissue processing cassettes, Histosette I

White

Price
889,00 kr
Log in for availability
1 * 500 Items
Product Details & Documents

Made of acetal polymer. With one-piece integral lid. To lock the cassette, just snap apart the lid, turn it round and lock it into the base of the cassette.

  • Writing area at 30° angle

Documents

Specifications
  • Colour:
    White
  • Seq:
    02
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.