Order Entry
Denmark
Orders LinkContactUsLinkComponent
Cyclohexanone ≥99.0% analytical reagent

Cyclohexanone ≥99.0% analytical reagent

Glass bottle, 1 L
Catalog # 23236.294
Flammable
Irritant
   Safer Human and Environmental Health
Supplier:   VWR Chemicals
Cyclohexanone ≥99.0% analytical reagent
Catalog # 23236.294
Supplier:  VWR Chemicals
   Safer Human and Environmental Health
CAS Number:  108-94-1
Cyclohexanone ≥99.0% analytical reagent
Cyclohexanone ≥99.0% analytical reagent

Glass bottle, 1 L

CAS Number:  108-94-1
Price
619,00 kr
Log in for availability
1 * 1 L
Product Details & Documents
Synonyms:  Pimelic Ketone
  • CAS Number:108-94-1
  • Formula:C₆H₁₀(=O)
  • MDL Number:MFCD00001625
  • Molecular Weight:98.14 g/mol
  • Merck Index:13,02755
  • Boiling Pt:155.6 °C (1013 hPa)
  • Melting Pt:–31 °C
  • Density:0.9478 g/cm³ (20 °C)
  • Flash Pt:43 °C
  • Storage Temperature:Ambient
  • EINECS:203-631-1
  • UN:1915
  • ADR:3,III
  • REACH:01-2119453616-35
Specifications

Specification Test Results

  • Assay:
    Min. 99.0 %
  • Acidity:
    Max. 0.005 meq/g
  • Boiling point:
    154.5 - 156.5 °C
  • Density (20/4):
    0.946 - 0.948
  • Refractive index (20°C):
    1.449 - 1.451
  • Viscosity (25°C):
    1.92 - 2.2 mPa·s
  • Cyclohexanol:
    Max. 0.5 %
  • Water:
    Max. 0.2 %

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Cat. No.:
    23236.294
  • Environmental certification:
    EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and after discussion
  • Environmentally Preferable:
    Safer Human and Environmental Health

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.