Cyclohexane ≥99%, GPR RECTAPUR®
Plastic jerrican, 5 L
Supplier: VWR Chemicals
Jump to:
Product Details & Documents
Synonyms: Hexahydrobenzene, Hexamethylene
- CAS Number:110-82-7
- Formula:C₆H₁₂
- MDL Number:MFCD00003814
- Molecular Weight:84.16 g/mol
- Merck Index:13,02752
- Boiling Pt:81 °C (1013 hPa)
- Melting Pt:7 °C
- Density:0.7785 g/cm³ (20 °C)
- Flash Pt:–18 °C
- Storage Temperature:Ambient
- EINECS:203-806-2
- UN:1145
- ADR:3,II
- REACH:01-2119463273-41
Documents
Certificates
Search by Lot Number1L Glass bottle for liquids DIN 45 (ZPACK13415101) 2,5L Glass bottle for liquids DIN 45 (ZPACK13415249) 225L Metal drum UN (ZPACE8010050) 30L Metal drum for liquids UN X (ZPACK12124125) 5L Plastic bottle for liquids DIN 45 UN X (ZPACK13951045)
Packaging Information
P20_VWRC23223.290_SPEC_EN.pdf P20_VWRC23223.324_SPEC_EN.pdf P20_VWRC23223.368_SPEC_EN.pdf P20_VWRC23223.461_SPEC_EN.pdf P20_VWRC23223.552_SPEC_EN.pdf
Product specification
More
VWR Chemicals Big Packs and Bulks
Product Literature
Specifications
Specification Test Results
- Assay:Min. 99 %
- IR Spectrum:Passes test
- Substances coloured by H2SO4:Passes test
- Boiling point:79 - 81 °C
- Density (20/4):0.778 - 0.779
- Free acidity:Max. 0.0002 meq/g
- Solidification point:5.5 - 6.5 °C
- Evaporation residue:Max. 50 ppm
Specifications
- Pack type:Plastic jerrican
- Pk:5 L
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



