Order Entry
Denmark
Orders LinkContactUsLinkComponent
pH Indicator solution, wide range pH 4 - 10

pH Indicator solution, wide range pH 4 - 10

Dropping bottle, marked, 100 mL
Catalog # HACH2329332
Supplier:   Hach
pH Indicator solution, wide range pH 4 - 10
Catalog # HACH2329332
Supplier:  Hach
CAS Number:  
pH Indicator solution, wide range pH 4 - 10
pH Indicator solution, wide range pH 4 - 10

Dropping bottle, marked, 100 mL

CAS Number:  
Price
321,00 kr
Log in for availability
1 * 100 mL
Product Details & Documents

Wide range of pH indicator solutions used for visual determination of pH, using test kits.

Method:10222

Specifications

Specifications

  • Pack type:
    Dropping bottle, marked
  • Pk:
    100 mL

Specifications